siRNA Details
| Gene Symbol: PGLS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012088.2 | NP_036220.1 | 4 | 643 | ||
| XM_011527897.2 | XP_011526199.1 | 4 | 1243 | |||
| GenBank mRNA | AF091091.1 | - | 3 | 743 | ||
| AJ243972.1 | - | 4 | 610 | |||
| BC014006.2 | - | 4 | 617 | 4053022 | ||
| BG764228.1 | - | 4 | 642 | |||
| GQ472229.1 | - | 4 | 617 | |||

PGLS
Genomic Map