siRNA Details
| Gene Symbol: QPCT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012413.3 | NP_036545.1 | 2, 3 | 383 | ||
| GenBank mRNA | AK290605.1 | - | 2, 3 | 365 | ||
| BC020456.1 | - | 2, 3 | 284 | |||
| BC036721.1 | - | 2, 3 | 262 | 5224222 | ||
| BC047756.2 | - | 2, 3 | 360 | 6058335 | ||
| BI755122.1 | - | 2, 3 | 418 | |||
| X67731.1 | - | 1, 2 | 65 | |||
| X71125.1 | - | 2, 3 | 272 | |||

QPCT
Genomic Map