siRNA Details
| Gene Symbol: QPCT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012413.3 | NP_036545.1 | 5 | 886 | ||
| GenBank mRNA | AK290605.1 | - | 5 | 868 | ||
| BC020456.1 | - | 5 | 787 | |||
| BC036721.1 | - | 5 | 765 | 5224222 | ||
| BC047756.2 | - | 5 | 863 | 6058335 | ||
| X67731.1 | - | 4 | 568 | |||
| X71125.1 | - | 5 | 775 | |||

QPCT
Genomic Map