siRNA Details
| Gene Symbol: BAMBI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012342.2 | NP_036474.1 | 3 | 983 | ||
| GenBank mRNA | AK223065.1 | - | 3 | 955 | ||
| AK290639.1 | - | 3 | 975 | |||
| BC019252.1 | - | 3 | 968 | 3503549 | ||
| BT006820.1 | - | 3 | 580 | |||
| U23070.1 | - | 3 | 952 | |||

BAMBI
Genomic Map