siRNA Details
| Gene Symbol: FAM19A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001082967.2 | NP_001076436.1 | 2 | 334 | ||
| NM_015381.6 | NP_056196.2 | 2 | 355 | |||
| GenBank mRNA | AF131851.1 | - | 2 | 300 | ||
| AK123443.1 | - | 3 | 294 | |||
| AK125256.1 | - | 2 | 355 | |||
| AY325118.1 | - | 2 | 329 | |||
| AY358847.1 | - | 3 | 230 | |||
| BC039396.1 | - | 2 | 313 | 5299791 | ||
| DB481230.1 | - | 2 | 334 | |||

FAM19A5
Genomic Map