siRNA Details
| Gene Symbol: DNAJB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135004.2 | NP_001128476.2 | 5 | 1367 | ||
| NM_001135005.2 | NP_001128477.1 | 4 | 1574 | |||
| NM_012266.5 | NP_036398.3 | 4 | 1222 | |||
| XM_005251428.3 | XP_005251485.1 | 4 | 1141 | |||
| XM_006716751.2 | XP_006716814.1 | 5 | 1231 | |||
| XM_011517842.2 | XP_011516144.1 | 5 | 1808 | |||
| GenBank mRNA | AF087870.1 | - | 5 | 1224 | ||
| AF088982.1 | - | 4 | 1125 | |||
| AK023253.1 | - | 4 | 1118 | |||
| AK299647.1 | - | 4 | 1574 | |||
| AK314671.1 | - | 4 | 1099 | |||

DNAJB5
Genomic Map