siRNA Details
| Gene Symbol: PARM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015393.3 | NP_056208.2 | 2 | 1004 | ||
| XM_011531833.1 | XP_011530135.1 | 3 | 1068 | |||
| GenBank mRNA | AK022311.1 | - | 2 | 963 | ||
| AL080121.1 | - | 2 | 920 | |||
| AY358777.1 | - | 2 | 949 | |||
| BC013294.2 | - | 2 | 933 | 3028327 | ||

PARM1
Genomic Map