siRNA Details
Gene Symbol: PDE4D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165899.1 | NP_001159371.1 | 1 | 100 | ||
XM_017009566.1 | XP_016865055.1 | 1 | 67 | |||
XM_017009568.1 | XP_016865057.1 | 1 | 27 | |||
XM_017009571.1 | XP_016865060.1 | 1 | 23 | |||
GenBank mRNA | AK307363.1 | - | 1 | 67 | ||
AY245866.1 | - | 1 | 65 | |||
DB230503.1 | - | 1 | 100 |
Gene Symbol: PART1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024617.1 | 1 | 283 | |||
NR_028508.1 | 1 | 64 | ||||
NR_028509.1 | 1 | 64 | ||||
GenBank mRNA | AF163475.1 | - | 1 | 64 | ||
AK312071.1 | - | 1 | 286 |
