siRNA Details
| Gene Symbol: NEPRO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319109.1 | NP_001306038.1 | 7 | 1339 | ||
| NM_001319110.1 | NP_001306039.1 | 6 | 1184 | |||
| NM_001319111.1 | NP_001306040.1 | 8 | 1153 | |||
| NM_001319112.1 | NP_001306041.1 | 8 | 1352 | |||
| NM_001319114.1 | NP_001306043.1 | 8 | 1398 | |||
| NM_001319115.1 | NP_001306044.1 | 8 | 1394 | |||
| NM_015412.3 | NP_056227.2 | 9 | 1524 | |||
| NR_134969.1 | 6 | 1277 | ||||
| XM_005247276.2 | XP_005247333.1 | 9 | 1491 | |||
| XM_011512635.2 | XP_011510937.1 | 9 | 1246 | |||
| XM_011512636.2 | XP_011510938.1 | 9 | 1250 | |||
| XM_011512638.2 | XP_011510940.1 | 8 | 1145 | |||
| XM_017006102.1 | XP_016861591.1 | 9 | 1513 | |||
| XM_017006103.1 | XP_016861592.1 | 8 | 1408 | |||
| GenBank mRNA | AK024325.1 | - | 9 | 1482 | ||
| AK295165.1 | - | 8 | 1283 | |||
| AK300811.1 | - | 8 | 1086 | |||
| AK301161.1 | - | 8 | 1312 | |||
| AK304022.1 | - | 3 | 612 | |||
| AK304083.1 | - | 8 | 1327 | |||
| BC014673.2 | - | 8 | 1982 | |||
| BC067743.1 | - | 9 | 1484 | 4812681 | ||
| CR749341.1 | - | 7 | 1340 | |||

NEPRO
Genomic Map