siRNA Details
| Gene Symbol: SPEF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015417.4 | NP_056232.2 | 2, 3 | 399 | ||
| XM_005260683.4 | XP_005260740.1 | 2, 3 | 410 | |||
| GenBank mRNA | AK313766.1 | - | 2, 3 | 412 | ||
| AL040853.1 | - | 1 | 13 | |||
| AL080154.1 | - | 2, 3 | 381 | |||
| BC022476.1 | - | 2, 3 | 358 | 4816764 | ||
| BI520334.1 | - | 2, 3 | 398 | |||
| DB030684.1 | - | 2, 3 | 412 | |||
| DB033051.1 | - | 2, 3 | 392 | |||
| EF560745.1 | - | 2, 3 | 397 | |||

SPEF1
Genomic Map