siRNA Details
| Gene Symbol: ZNF473 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017026574.1 | XP_016882063.1 | 3 | 3326 | ||
| GenBank mRNA | AB032967.1 | - | 5 | 2083 | ||
| AF454744.1 | - | 4 | 2038 | |||
| AK054671.1 | - | 1 | 1225 | |||
| AK292452.1 | - | 5 | 1976 | |||
| AK302293.1 | - | 4 | 1933 | |||
| AL080143.1 | - | 1 | 391 | |||
| BC018612.1 | - | 5 | 2006 | 4634056 | ||
| BX648093.1 | - | 5 | 2127 | |||

ZNF473
Genomic Map