siRNA Details
| Gene Symbol: PAMR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001991.2 | NP_001001991.1 | 10 | 1994 | ||
| NM_001282675.1 | NP_001269604.1 | 12 | 1984 | |||
| NM_001282676.1 | NP_001269605.1 | 8 | 1661 | |||
| NM_015430.3 | NP_056245.2 | 11 | 2045 | |||
| GenBank mRNA | AF370388.1 | - | 9 | 1483 | ||
| AK027841.1 | - | 11 | 1645 | |||
| AK297092.1 | - | 12 | 1985 | |||
| AK300185.1 | - | 8 | 1246 | |||
| AK316188.1 | - | 8 | 1233 | |||
| AL832391.1 | - | 10 | 1560 | |||
| AY358346.1 | - | 10 | 1573 | |||
| BC089434.1 | - | 11 | 1637 | 30558617 | ||
| BX640676.1 | - | 10 | 1994 | |||

PAMR1
Genomic Map