siRNA Details
| Gene Symbol: PAMR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001991.2 | NP_001001991.1 | 6 | 1205 | ||
| NM_001282675.1 | NP_001269604.1 | 8 | 1195 | |||
| NM_001282676.1 | NP_001269605.1 | 4 | 872 | |||
| NM_015430.3 | NP_056245.2 | 6 | 1205 | |||
| GenBank mRNA | AF370388.1 | - | 5 | 693 | ||
| AK297092.1 | - | 8 | 1196 | |||
| AK300185.1 | - | 4 | 457 | |||
| AK316188.1 | - | 4 | 444 | |||
| AL832391.1 | - | 6 | 771 | |||
| AY358346.1 | - | 6 | 784 | |||
| BC089434.1 | - | 6 | 797 | 30558617 | ||
| BX640676.1 | - | 6 | 1205 | |||

PAMR1
Genomic Map