siRNA Details
| Gene Symbol: CCDC28A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015439.2 | NP_056254.1 | 4 | 840 | ||
| GenBank mRNA | AJ420568.1 | - | 4 | 591 | ||
| AK300672.1 | - | 4 | 591 | |||
| AL050197.1 | - | 4 | 590 | |||
| AY167571.1 | - | 4 | 858 | |||
| BC004464.1 | - | 4 | 575 | 2820737 | ||
| BC013019.2 | - | 4 | 572 | |||
| BC080566.1 | - | 4 | 574 | |||
| BC108717.1 | - | 4 | 763 | 6068596 | ||

CCDC28A
Genomic Map