siRNA Details
| Gene Symbol: SEC31B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015490.3 | NP_056305.1 | 15 | 1936 | ||
| GenBank mRNA | AF274863.1 | - | 15 | 1932 | ||
| AK128343.1 | - | 9 | 2070 | |||
| AL080141.1 | - | 8 | 1044 | |||
| BC034946.1 | - | 15 | 2492 | |||
| BC044569.1 | - | 15 | 2485 | |||
| BC053949.1 | - | 15 | 2485 | |||
| BC071580.1 | - | 15 | 2412 | |||
| BC144313.1 | - | 14 | 1874 | 9052833 | ||
| BX640914.1 | - | 15 | 1833 | |||

SEC31B
Genomic Map