siRNA Details
| Gene Symbol: UNC50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014044.5 | NP_054763.2 | 3 | 477 | ||
| XM_005263913.4 | XP_005263970.1 | 3 | 573 | |||
| XM_006712403.3 | XP_006712466.1 | 3 | 573 | |||
| XM_006712404.2 | XP_006712467.1 | 3 | 487 | |||
| GenBank mRNA | AF077038.1 | - | 3 | 429 | ||
| AK222599.1 | - | 3 | 443 | |||
| AL080115.1 | - | 3 | 481 | |||
| AY017215.1 | - | 3 | 445 | |||
| AY251534.1 | - | 2 | 338 | |||
| BC017019.1 | - | 3 | 610 | 3888349 | ||
| BC025992.2 | - | 2 | 1505 | 4830706 | ||
| BC039078.1 | - | 2 | 1481 | 5166015 | ||
| BC091481.1 | - | 3 | 467 | 30337350 | ||
| CR533464.1 | - | 2 | 325 | |||
| KU178586.1 | - | 2 | 325 | |||
| KU178587.1 | - | 2 | 325 | |||

UNC50
Genomic Map