siRNA Details
| Gene Symbol: EGFL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167890.1 | NP_001161362.1 | 5, 6 | 796 | ||
| NM_015507.3 | NP_056322.2 | 5, 6 | 796 | |||
| GenBank mRNA | AF186084.1 | - | 5, 6 | 654 | ||
| AF193055.1 | - | 5, 6 | 721 | |||
| AJ245671.1 | - | 5, 6 | 749 | |||
| AK075214.1 | - | 5, 6 | 766 | |||
| AK300137.1 | - | 5, 6 | 532 | |||
| AK300236.1 | - | 5, 6 | 766 | |||
| AK315018.1 | - | 5, 6 | 634 | |||
| AL117610.1 | - | 5, 6 | 526 | |||
| AY358333.1 | - | 5, 6 | 643 | |||
| BC038587.1 | - | 5, 6 | 605 | 5767603 | ||

EGFL6
Genomic Map