siRNA Details
| Gene Symbol: LTN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320766.1 | NP_001307695.1 | 20 | 3843 | ||
| NM_015565.2 | NP_056380.2 | 21 | 3882 | |||
| XM_017028316.1 | XP_016883805.1 | 20 | 3562 | |||
| XM_017028317.1 | XP_016883806.1 | 11 | 2157 | |||
| GenBank mRNA | AB018257.2 | - | 21 | 3773 | ||
| BC140790.1 | - | 21 | 3795 | 9021658 | ||
| BC150284.1 | - | 21 | 3773 | 8860138 | ||
| CN361592.1 | - | 5 | 545 | |||

LTN1
Genomic Map