siRNA Details
| Gene Symbol: CNTNAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014141.5 | NP_054860.1 | 3 | 840 | ||
| XM_017011950.1 | XP_016867439.1 | 3 | 1166 | |||
| GenBank mRNA | AB020675.1 | - | 3 | 349 | ||
| AF319045.1 | - | 3 | 464 | |||
| AK315113.1 | - | 3 | 429 | |||
| BC093780.1 | - | 3 | 367 | 7939625 | ||
| BC113373.1 | - | 3 | 367 | 8322425 | ||
| KJ534811.1 | - | 2 | 213 | |||
| KJ535069.1 | - | 324 | ||||

CNTNAP2
Genomic Map