siRNA Details
| Gene Symbol: ANKRD17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286771.2 | NP_001273700.1 | 6 | 927 | ||
| NM_015574.1 | NP_056389.1 | 6 | 1309 | |||
| NM_032217.4 | NP_115593.3 | 6 | 1309 | |||
| NM_198889.2 | NP_942592.1 | 6 | 1309 | |||
| XM_005265671.3 | XP_005265728.1 | 6 | 1653 | |||
| XM_005265672.3 | XP_005265729.1 | 6 | 1653 | |||
| XM_005265673.3 | XP_005265730.1 | 6 | 1653 | |||
| XM_017008011.1 | XP_016863500.1 | 6 | 884 | |||
| XM_017008012.1 | XP_016863501.1 | 6 | 884 | |||
| XM_017008013.1 | XP_016863502.1 | 6 | 884 | |||
| XM_017008014.1 | XP_016863503.1 | 6 | 1653 | |||
| XM_017008015.1 | XP_016863504.1 | 6 | 884 | |||
| XM_017008016.1 | XP_016863505.1 | 6 | 1653 | |||
| XM_017008017.1 | XP_016863506.1 | 6 | 884 | |||
| GenBank mRNA | AB014597.2 | - | 6 | 833 | ||
| AF308285.1 | - | 6 | 1296 | |||
| AK300302.1 | - | 6 | 883 | |||
| AK307581.1 | - | 6 | 1309 | |||
| AL833287.1 | - | 3 | 486 | |||
| BC004173.2 | - | 6 | 1290 | |||
| BC007747.2 | - | 4 | 639 | |||
| BC043394.1 | - | 6 | 1278 | |||
| BX648035.1 | - | 6 | 882 | |||

ANKRD17
Genomic Map