siRNA Details
| Gene Symbol: KIF1BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015634.3 | NP_056449.1 | 6, 7 | 1097 | ||
| XM_017016067.1 | XP_016871556.1 | 3, 4 | 1621 | |||
| GenBank mRNA | AB033105.1 | - | 6, 7 | 1020 | ||
| AK291343.1 | - | 6, 7 | 1002 | |||
| AL050190.1 | - | 1, 2 | 48 | |||
| BC006427.1 | - | 6, 7 | 1020 | |||
| BC012180.1 | - | 6, 7 | 994 | 4550085 | ||

KIF1BP
Genomic Map