siRNA Details
| Gene Symbol: C10orf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015652.2 | NP_056467.2 | 1 | 1901 | ||
| GenBank mRNA | AK023084.1 | - | 1 | 1090 | ||
| AK126886.1 | - | 1 | 1954 | |||
| BC024315.1 | - | 1 | 1901 | 3903728 | ||
| BC031482.1 | - | 1 | 1298 | |||

C10orf12
Genomic Map