siRNA Details
| Gene Symbol: ZNF337 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290261.1 | NP_001277190.1 | 2, 3 | 678 | ||
| NM_015655.3 | NP_056470.1 | 3, 4 | 351 | |||
| XM_006723558.3 | XP_006723621.1 | 3, 4 | 1354 | |||
| XM_011529219.2 | XP_011527521.1 | 3, 4 | 474 | |||
| GenBank mRNA | AK056184.1 | - | 3, 4 | 263 | ||
| AK125455.1 | - | 2, 3 | 681 | |||
| AK291235.1 | - | 3, 4 | 288 | |||
| BC021298.2 | - | 3, 4 | 280 | 4551314 | ||
| BP201424.1 | - | 3, 4 | 351 | |||

ZNF337
Genomic Map