siRNA Details
| Gene Symbol: GMEB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012384.4 | NP_036516.1 | 8, 9 | 1029 | ||
| XM_005260202.4 | XP_005260259.1 | 8, 9 | 1240 | |||
| XM_006723776.2 | XP_006723839.1 | 6, 7 | 681 | |||
| XM_011528779.2 | XP_011527081.1 | 6, 7 | 745 | |||
| GenBank mRNA | AB033095.1 | - | 8, 9 | 906 | ||
| AF173867.1 | - | 7, 8 | 1305 | |||
| AL133646.1 | - | 6, 7 | 753 | |||
| BC036305.1 | - | 8, 9 | 956 | 5166938 | ||
| BQ954281.1 | - | 7, 8 | 731 | |||

GMEB2
Genomic Map