siRNA Details
| Gene Symbol: FBXL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193534.1 | NP_001180463.1 | 9 | 1527 | ||
| NM_001193535.1 | NP_001180464.1 | 9 | 1479 | |||
| NM_012161.3 | NP_036293.1 | 9 | 1530 | |||
| NR_036464.1 | 10 | 1634 | ||||
| XM_006713959.3 | XP_006714022.1 | 10 | 1626 | |||
| XM_011513831.2 | XP_011512133.1 | 9 | 1312 | |||
| XM_011513832.2 | XP_011512134.1 | 9 | 1459 | |||
| XM_011513833.2 | XP_011512135.1 | 8 | 1270 | |||
| XM_017008019.1 | XP_016863508.1 | 11 | 2397 | |||
| GenBank mRNA | AF052097.1 | - | 1 | 89 | ||
| AF142481.1 | - | 9 | 1452 | |||
| AF157323.1 | - | 9 | 1391 | |||
| AF174591.1 | - | 6 | 937 | |||
| AF176700.1 | - | 9 | 1414 | |||
| AF199420.1 | - | 9 | 1416 | |||
| AK000153.1 | - | 8 | 1240 | |||
| AK295510.1 | - | 9 | 1449 | |||
| AK310860.1 | - | 9 | 1459 | |||
| AL832808.1 | - | 7 | 1166 | |||
| BC012320.1 | - | 10 | 1563 | |||
| BC030656.1 | - | 9 | 1501 | 4837080 | ||
| BX537957.1 | - | 9 | 1425 | |||

FBXL5
Genomic Map