siRNA Details
| Gene Symbol: OR2M4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017504.1 | NP_059974.1 | 1 | 596 | ||
| GenBank mRNA | X64992.1 | - | 1 | 222 | ||
| X89666.1 | - | 1 | 222 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR2M4| Gene Symbol: OR2M4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017504.1 | NP_059974.1 | 1 | 596 | ||
| GenBank mRNA | X64992.1 | - | 1 | 222 | ||
| X89666.1 | - | 1 | 222 | |||