siRNA Details
| Gene Symbol: KLHL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001257194.1 | NP_001244123.1 | 8 | 1459 | ||
| NM_001257195.1 | NP_001244124.1 | 6 | 1035 | |||
| NM_017415.2 | NP_059111.2 | 8 | 1284 | |||
| GenBank mRNA | AB032955.1 | - | 8 | 1054 | ||
| AF208068.1 | - | 8 | 968 | |||
| AF208069.1 | - | 6 | 1072 | |||
| AF208070.1 | - | 8 | 1459 | |||
| AK300172.1 | - | 3 | 465 | |||
| AK303916.1 | - | 6 | 1030 | |||
| AK310568.1 | - | 7 | 954 | |||
| AK311594.1 | - | 3 | 430 | |||
| AK314707.1 | - | 8 | 1025 | |||
| BC034035.1 | - | 8 | 1283 | 4817650 | ||

KLHL3
Genomic Map