siRNA Details
| Gene Symbol: FBXO5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142522.1 | NP_001135994.1 | 2 | 774 | ||
| NM_012177.3 | NP_036309.1 | 2 | 614 | |||
| GenBank mRNA | AF129535.1 | - | 2 | 566 | ||
| AJ420499.1 | - | 2 | 634 | |||
| AK026197.1 | - | 3 | 523 | |||
| AK055221.1 | - | 2 | 748 | |||
| AK314456.1 | - | 2 | 596 | |||
| AY079515.1 | - | 2 | 505 | |||
| BC018905.1 | - | 2 | 556 | 3960052 | ||

FBXO5
Genomic Map