siRNA Details
| Gene Symbol: OR8G1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002905.1 | NP_001002905.1 | 1 | 739 | ||
| NR_045681.1 | 1 | 739 | ||||
| GenBank mRNA | X89672.1 | - | 1 | 365 | ||
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 OR8G1
OR8G1| Gene Symbol: OR8G1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002905.1 | NP_001002905.1 | 1 | 739 | ||
| NR_045681.1 | 1 | 739 | ||||
| GenBank mRNA | X89672.1 | - | 1 | 365 | ||
 Back To Top
 Back To Top