siRNA Details
| Gene Symbol: TSPAN16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282509.1 | NP_001269438.1 | 5, 6 | 765 | ||
| NM_001282510.1 | NP_001269439.1 | 4, 5 | 690 | |||
| NM_012466.3 | NP_036598.1 | 5, 6 | 765 | |||
| NR_104199.1 | 5, 6 | 798 | ||||
| XM_011527902.1 | XP_011526204.1 | 4, 5 | 652 | |||
| XM_011527903.2 | XP_011526205.1 | 5, 6 | 696 | |||
| GenBank mRNA | AW169944.1 | - | 2, 3 | 260 | ||
| BC029908.1 | - | 5, 6 | 768 | 5168677 | ||
| BM560276.1 | - | 5, 6 | 752 | |||

TSPAN16
Genomic Map