siRNA Details
| Gene Symbol: OR10H3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013938.1 | NP_039226.1 | 1 | 795 | ||
| GenBank mRNA | BC106707.2 | - | 1 | 795 | 40004023 | |
| BC113844.1 | - | 1 | 795 | 40004027 | ||
More Information
Set Membership: |
|

OR10H3
Genomic Map