siRNA Details
| Gene Symbol: OSTF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012383.4 | NP_036515.4 | 2, 3 | 294 | ||
| XM_006717053.3 | XP_006717116.1 | 2, 3 | 303 | |||
| XM_011518525.2 | XP_011516827.1 | 2, 3 | 305 | |||
| XM_017014622.1 | XP_016870111.1 | 2, 3 | 295 | |||
| GenBank mRNA | AK222596.1 | - | 2, 3 | 238 | ||
| AK291511.1 | - | 2, 3 | 262 | |||
| BC007459.1 | - | 2, 3 | 200 | 4052054 | ||
| BP312980.1 | - | 2, 3 | 294 | |||
| U63717.1 | - | 2, 3 | 232 | |||

OSTF1
Genomic Map