siRNA Details
| Gene Symbol: MYEOV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293291.1 | NP_001280220.1 | 3 | 829 | ||
| NM_001293294.1 | NP_001280223.1 | 2 | 925 | |||
| NM_001293296.1 | NP_001280225.1 | 2 | 757 | |||
| NM_001300923.1 | NP_001287852.1 | 3 | 661 | |||
| NM_001300924.1 | NP_001287853.1 | 2 | 566 | |||
| NM_138768.3 | NP_620123.2 | 3 | 735 | |||
| GenBank mRNA | AJ223366.1 | - | 2 | 903 | ||
| AK026148.1 | - | 2 | 737 | |||
| AK290121.1 | - | 3 | 811 | |||
| BC011815.2 | - | 3 | 693 | 3163949 | ||
| BM785815.1 | - | 3 | 487 | |||

MYEOV
Genomic Map