siRNA Details
Gene Symbol: OR7C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012377.1 | NP_036509.1 | 1 | 640 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
