siRNA Details
| Gene Symbol: OR2J2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030905.2 | NP_112167.2 | 1 | 812 | ||
| GenBank mRNA | BC069121.1 | - | 1 | 710 | 7216975 | |
| BC108896.1 | - | 1 | 713 | 40003949 | ||
| BC108897.1 | - | 1 | 710 | 40003955 | ||
| BC108898.1 | - | 1 | 710 | 40003956 | ||
| BC108899.1 | - | 1 | 710 | 40003957 | ||

OR2J2
Genomic Map