siRNA Details
| Gene Symbol: OPLAH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017570.4 | NP_060040.1 | 14 | 1967 | ||
| XM_011516960.1 | XP_011515262.1 | 14 | 2243 | |||
| GenBank mRNA | AB122018.1 | - | 13 | 1863 | ||
| AL096750.1 | - | 6 | 952 | |||
| BC036944.1 | - | 9 | 1154 | |||
| BC142672.1 | - | 14 | 1967 | 40148181 | ||
| BC150206.1 | - | 14 | 1945 | 40148883 | ||

OPLAH
Genomic Map