siRNA Details
| Gene Symbol: RNF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014372.4 | NP_055187.1 | 3 | 848 | ||
| GenBank mRNA | AB024703.1 | - | 3 | 491 | ||
| AF151881.1 | - | 3 | 491 | |||
| AK313140.1 | - | 3 | 610 | |||
| BC020964.2 | - | 3 | 443 | 3917224 | ||
| BC047654.1 | - | 3 | 816 | 5261966 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_013876, AK178138, AF220206, AB024427, BC010299, and others.

RNF11
Genomic Map