siRNA Details
| Gene Symbol: MLH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040108.1 | NP_001035197.1 | 2 | 3165 | ||
| NM_014381.2 | NP_055196.2 | 2 | 3165 | |||
| XM_005267532.4 | XP_005267589.1 | 2 | 3165 | |||
| XM_005267533.4 | XP_005267590.1 | 2 | 3165 | |||
| XM_005267534.2 | XP_005267591.1 | 2 | 3165 | |||
| XM_006720116.3 | XP_006720179.1 | 2 | 3077 | |||
| XM_011536646.2 | XP_011534948.1 | 2 | 3165 | |||
| XM_011536648.2 | XP_011534950.1 | 2 | 3165 | |||
| XM_017021219.1 | XP_016876708.1 | 2 | 3091 | |||
| GenBank mRNA | AB039667.1 | - | 1 | 2949 | ||
| AF195657.1 | - | 2 | 3063 | |||
| AL833875.1 | - | 1 | 995 | |||
| BC112167.1 | - | 1 | 2985 | 8327635 | ||

MLH3
Genomic Map