siRNA Details
| Gene Symbol: ZBTB11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014415.3 | NP_055230.2 | 4 | 1608 | ||
| XM_011512689.2 | XP_011510991.1 | 4 | 1316 | |||
| XM_011512690.2 | XP_011510992.1 | 2 | 2993 | |||
| GenBank mRNA | AB208862.1 | - | 1 | 321 | ||
| AK312859.1 | - | 4 | 1600 | |||
| BC111700.1 | - | 4 | 1515 | 40038346 | ||
| U69274.1 | - | 4 | 1539 | |||

ZBTB11
Genomic Map