siRNA Details
| Gene Symbol: FAM155B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015686.2 | NP_056501.2 | 1 | 869 | ||
| XM_011530908.2 | XP_011529210.1 | 1 | 900 | |||
| XM_011530909.2 | XP_011529211.1 | 1 | 900 | |||
| GenBank mRNA | AF087142.1 | - | 1 | 868 | ||
| BC136518.1 | - | 1 | 856 | 9020508 | ||
| BC136519.1 | - | 1 | 856 | 9020509 | ||

FAM155B
Genomic Map