siRNA Details
| Gene Symbol: DKK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014421.2 | NP_055236.1 | 1, 2 | 936 | ||
| GenBank mRNA | AB033208.1 | - | 1, 2 | 453 | ||
| AB033941.1 | - | 1, 2 | 453 | |||
| AF177395.1 | - | 1, 2 | 948 | |||
| AK309466.1 | - | 2, 3 | 165 | |||
| AK312695.1 | - | 1, 2 | 462 | |||
| AY358414.1 | - | 1, 2 | 441 | |||
| BC075077.2 | - | 1, 2 | 265 | 30915382 | ||
| BC075078.2 | - | 1, 2 | 265 | 30915595 | ||
| BC126328.1 | - | 1, 2 | 537 | 8992044 | ||
| BC126330.1 | - | 1, 2 | 537 | 8992046 | ||

DKK2
Genomic Map