siRNA Details
| Gene Symbol: SMC1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291501.1 | NP_001278430.1 | 18 | 2840 | ||
| NM_148674.4 | NP_683515.4 | 18 | 2840 | |||
| XM_011530144.2 | XP_011528446.1 | 18 | 2701 | |||
| XM_011530145.2 | XP_011528447.1 | 18 | 2892 | |||
| GenBank mRNA | AJ504806.2 | - | 18 | 2840 | ||
| AK125736.1 | - | 5 | 599 | |||
| BC126208.1 | - | 18 | 2820 | 8991924 | ||

SMC1B
Genomic Map