siRNA Details
| Gene Symbol: STK36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243313.1 | NP_001230242.1 | 6 | 948 | ||
| NM_015690.4 | NP_056505.2 | 6 | 948 | |||
| XM_005246464.1 | XP_005246521.1 | 6 | 836 | |||
| XM_011510959.1 | XP_011509261.1 | 6 | 948 | |||
| XM_011510960.1 | XP_011509262.1 | 6 | 889 | |||
| XM_017003804.1 | XP_016859293.1 | 6 | 836 | |||
| XM_017003805.1 | XP_016859294.1 | 5 | 616 | |||
| GenBank mRNA | AB033104.1 | - | 6 | 905 | ||
| AF200815.1 | - | 5 | 669 | |||
| BC026158.2 | - | 6 | 888 | 4839214 | ||

STK36
Genomic Map