siRNA Details
| Gene Symbol: RSPH14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014433.2 | NP_055248.1 | 7 | 1181 | ||
| XM_011530149.2 | XP_011528451.1 | 8 | 1370 | |||
| XM_011530150.1 | XP_011528452.1 | 8 | 1353 | |||
| XM_011530151.2 | XP_011528453.1 | 8 | 1516 | |||
| XM_011530152.2 | XP_011528454.1 | 8 | 1444 | |||
| XM_011530154.2 | XP_011528456.1 | 7 | 1059 | |||
| XM_011530155.2 | XP_011528457.1 | 5 | 1046 | |||
| XM_017028774.1 | XP_016884263.1 | 8 | 1339 | |||
| GenBank mRNA | AF133587.1 | - | 7 | 1181 | ||
| BC008986.1 | - | 7 | 1149 | 4339021 | ||
| CR456567.1 | - | 6 | 1081 | |||

RSPH14
Genomic Map