siRNA Details
| Gene Symbol: RSPH14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014433.2 | NP_055248.1 | 5, 6 | 801 | ||
| XM_011530149.2 | XP_011528451.1 | 5, 6 | 840 | |||
| XM_011530150.1 | XP_011528452.1 | 5, 6 | 840 | |||
| XM_011530151.2 | XP_011528453.1 | 5, 6 | 1003 | |||
| XM_011530152.2 | XP_011528454.1 | 5, 6 | 931 | |||
| XM_011530154.2 | XP_011528456.1 | 4, 5 | 546 | |||
| XM_011530155.2 | XP_011528457.1 | 2, 3 | 533 | |||
| XM_017028774.1 | XP_016884263.1 | 5, 6 | 826 | |||
| GenBank mRNA | AF133587.1 | - | 5, 6 | 801 | ||
| BC008986.1 | - | 5, 6 | 769 | 4339021 | ||
| CR456567.1 | - | 4, 5 | 701 | |||

RSPH14
Genomic Map