siRNA Details
| Gene Symbol: SLC39A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271957.1 | NP_001258886.1 | 4 | 1183 | ||
| NM_001271958.1 | NP_001258887.1 | 4 | 1020 | |||
| NM_001271959.1 | NP_001258888.1 | 4 | 1189 | |||
| NM_001271960.1 | NP_001258889.1 | 5 | 1081 | |||
| NM_001271961.1 | NP_001258890.1 | 3 | 889 | |||
| NM_014437.4 | NP_055252.2 | 5 | 1327 | |||
| GenBank mRNA | AF132942.1 | - | 3 | 877 | ||
| AF151829.1 | - | 4 | 898 | |||
| AJ243649.1 | - | 4 | 914 | |||
| AJ271671.1 | - | 4 | 928 | |||
| AK074943.1 | - | 4 | 1137 | |||
| AK293389.1 | - | 3 | 797 | |||
| AK297983.1 | - | 5 | 895 | |||
| AK300244.1 | - | 5 | 887 | |||
| BC002563.2 | - | 4 | 902 | 3138575 | ||
| BC003152.2 | - | 3 | 874 | 2958651 | ||
| BC007886.2 | - | 4 | 899 | 4122086 | ||
| BC014303.2 | - | 5 | 1307 | 3942586 | ||
| CR457217.1 | - | 3 | 841 | |||

SLC39A1
Genomic Map