siRNA Details
| Gene Symbol: VPS4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013245.2 | NP_037377.1 | 3 | 336 | ||
| GenBank mRNA | AF112215.1 | - | 3 | 291 | ||
| AF132747.1 | - | 3 | 344 | |||
| AF155740.1 | - | 2 | 194 | |||
| AF159063.1 | - | 3 | 336 | |||
| AF255952.1 | - | 3 | 336 | |||
| AK315026.1 | - | 3 | 364 | |||
| BC047932.1 | - | 3 | 314 | 5753632 | ||

VPS4A
Genomic Map