siRNA Details
Gene Symbol: SULT1C4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006588.3 | NP_006579.2 | 2, 3 | 716 | ||
XM_017003807.1 | XP_016859296.1 | 2, 3 | 258 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SULT1C4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006588.3 | NP_006579.2 | 2, 3 | 716 | ||
XM_017003807.1 | XP_016859296.1 | 2, 3 | 258 |