siRNA Details
Gene Symbol: KLHL20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014458.3 | NP_055273.2 | 3, 4 | 767 | ||
XM_005245093.3 | XP_005245150.1 | 4, 5 | 916 | |||
XM_011509420.2 | XP_011507722.1 | 3, 4 | 796 | |||
XM_017001052.1 | XP_016856541.1 | 3, 4 | 1046 | |||
XM_017001053.1 | XP_016856542.1 | 3, 4 | 1046 | |||
XM_017001054.1 | XP_016856543.1 | 4, 5 | 916 | |||
GenBank mRNA | AB026190.1 | - | 3, 4 | 708 | ||
AJ844466.1 | - | 2, 3 | 588 | |||
AK001430.1 | - | 3, 4 | 724 | |||
AK308048.1 | - | 2, 3 | 685 | |||
BC063418.1 | - | 3, 4 | 697 | 5221643 |
