siRNA Details
| Gene Symbol: RND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014470.3 | NP_055285.1 | 1 | 142 | ||
| GenBank mRNA | AB040147.1 | - | 1 | 120 | ||
| AF498967.1 | - | 1 | 39 | |||
| AK292762.1 | - | 1 | 142 | |||
| BC026356.1 | - | 1 | 142 | 4793442 | ||
| Y07923.1 | - | 1 | 39 | |||

RND1
Genomic Map